Methyl-2-(bromomethyl)-6-nitrobenzoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-(bromomethyl)-6-nitrobenzoate

A256095-250MG A256095-1G
103812-242EA 242 USD
103812-242 103852-104 76815-858 77091-582
Methyl-2-(bromomethyl)-6-nitrobenzoate 98%
Methyl 2-(bromomethyl)-6-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MW: 274.0700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114314
CAS Number: 61940-21-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR