Methyl-2-(bromomethyl)-3-chlorobenzoate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-(bromomethyl)-3-chlorobenzoate

A381153-1G A381153-250MG
103826-690EA 67 USD
103826-690 103837-724 77306-922 77890-785
Methyl-2-(bromomethyl)-3-chlorobenzoate 97%
Methyl 2-(bromomethyl)-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.5156 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114322
CAS Number: 188187-03-3
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR