Methyl-2-(benzylamino)acetate 98%

Supplier: Ambeed
Danger

Synonyms: N-Benzylglycine methyl ester, Methyl 2-(benzylamino)acetate

A606442-10G A606442-250MG A606442-1G A606442-25G A606442-5G
103829-002EA 9 USD
103829-002 103840-142 76806-036 76806-038 77307-924 77307-926 77307-928
Methyl-2-(benzylamino)acetate 98%
Methyl-2-(benzylamino)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03247461
CAS Number: 53386-64-4
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR