Methyl-2-(aminomethyl)pyridine-4-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(aminomethyl)pyridine-4-carboxylate hydrochloride

A256559-100MG A256559-250MG A256559-1G A256559-5G
103812-274EA 88 USD
103812-274 103852-138 76976-836 76976-838 77304-682 77304-684
Methyl-2-(aminomethyl)pyridine-4-carboxylate hydrochloride 97%
Methyl-2-(aminomethyl)pyridine-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂
MW: 202.6400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878665
CAS Number: 1072438-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR