Methyl 2-(aminomethyl)nicotinate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A105937-100MG
A105937-250MG
A105937-1G
103796-974EA
87
USD
103796-974
103822-722
77148-468
77148-470
77309-584
Methyl 2-(aminomethyl)nicotinate hydrochloride 97%
Methyl 2-(aminomethyl)nicotinate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₁ClN₂O₂ MW: 202.64 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18803301 CAS Number: 151509-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...