Methyl 2-(6-chloropyridin-3-yl)acetate 97%

Supplier: Ambeed
Warning

A143936-100G A143936-250MG A143936-1G A143936-25G
103803-352EA 8 USD
103803-352 76984-318 76984-320 77316-658 77316-660
Methyl 2-(6-chloropyridin-3-yl)acetate 97%
Methyl 2-(6-chloropyridin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15526721
CAS Number: 717106-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR