Methyl 2-(6-chloropyridin-2-yl)acetate 97%

Supplier: Ambeed
Warning

A315043-250MG A315043-1G
103825-054EA 133 USD
103825-054 103835-980 76977-970 77306-122
Methyl 2-(6-chloropyridin-2-yl)acetate 97%
Methyl 2-(6-chloropyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834576
CAS Number: 161807-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR