Methyl 2-(5-nitropyridin-2-yl)acetate 97%

Supplier: Ambeed
Warning

A162703-100MG A162703-250MG A162703-1G A162703-5G
103805-650EA 32 USD
103805-650 103843-900 76824-126 76824-128 77319-972 77319-974
Methyl 2-(5-nitropyridin-2-yl)acetate 97%
Methyl 2-(5-nitropyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15526724
CAS Number: 292600-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR