Methyl 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate 97%

Supplier: Ambeed
Warning

A214681-100MG A214681-250MG A214681-1G A214681-25G A214681-5G
103818-426EA 41 USD
103818-426 103849-130 77152-572 77152-574 77303-364 77303-366 77303-368
Methyl 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate 97%
Methyl 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00218549
CAS Number: 7588-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR