(4-Hydroxyphenyl)acetic acid methyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-hydroxyphenylacetate, Methyl (4-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate
A166602-100G
A166602-500G
A166602-25G
A166602-10G
103808-974EA
172
USD
103808-974
77247-856
77247-858
77320-616
77746-124
(4-Hydroxyphenyl)acetic acid methyl ester 97%
(4-Hydroxyphenyl)acetic acid methyl ester
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 162…163 °C (760 mm Hg) Melting Pt: 55…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002387 CAS Number: 14199-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...