Methyl 2-(4-formylphenyl)acetate 98%
Supplier: Ambeed
|
Warning
|
A197021-10G
A197021-250MG
A197021-1G
A197021-25G
A197021-5G
103816-886EA
16
USD
103816-886
103847-502
76785-156
76785-158
77085-480
77085-482
77085-484
Methyl 2-(4-formylphenyl)acetate 98%
Methyl 2-(4-formylphenyl)acetate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₃ MW: 178.185 g/mol Boiling Pt: 285 °C (760 mm Hg) Flash Pt: 124 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD17018734 CAS Number: 96524-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...