Methyl (4-fluorophenyl)acetate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-(4-fluorophenyl)acetate
A273401-100G
A273401-10G
A273401-25G
A273401-5G
A273401-1G
103813-142EA
9
USD
103813-142
103853-062
76723-088
76723-090
77092-860
77092-862
77305-130
Methyl (4-fluorophenyl)acetate 97%
Methyl (4-fluorophenyl)acetate
Precaución: for Research Use Only
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Boiling Pt: 211 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00800608 CAS Number: 34837-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...