Methyl 2-(4-fluoro-3-nitrophenyl)acetate 95%

Supplier: Ambeed
Warning

A131024-100MG A131024-250MG A131024-1G
103801-512EA 23 USD
103801-512 76972-052 76972-054 77313-988
Methyl 2-(4-fluoro-3-nitrophenyl)acetate 95%
Methyl 2-(4-fluoro-3-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈FNO₄
MW: 213.16 g/mol
Boiling Pt: 320.5 °C (760 mm Hg)
Flash Pt: 147.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09909451
CAS Number: 226888-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR