Methyl 2-(4-cyanophenyl)-2-methylpropanoate 95%

Supplier: Ambeed
Warning

A142508-100MG A142508-250MG
103803-172EA 506 USD
103803-172 77010-418 77010-420
Methyl 2-(4-cyanophenyl)-2-methylpropanoate 95%
Methyl 2-(4-cyanophenyl)-2-methylpropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD23097041
CAS Number: 444807-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR