Methyl-2-(4-bromophenyl)-2,2-dimethylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-bromophenyl)-2,2-dimethylacetate

A121826-100G A121826-25G
103800-140EA 6 USD
103800-140 76995-842 76995-844
Methyl-2-(4-bromophenyl)-2,2-dimethylacetate 98%
Methyl-2-(4-bromophenyl)-2,2-dimethylacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.1237 g/mol
Boiling Pt: 290.3 °C (760 mm Hg)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD02683357
CAS Number: 154825-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR