Methyl-4-bromo-2-fluorophenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-bromo-2-fluorophenyl)acetate

A142614-100G A142614-100MG A142614-10G A142614-250MG A142614-25G A142614-1G A142614-5G
103803-194EA 10 USD
103803-194 76811-836 76811-838 76811-840 76811-842 76811-844 77316-428 77316-430
Methyl-4-bromo-2-fluorophenylacetate 98%
Methyl-4-bromo-2-fluorophenylacetate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.063 g/mol
Boiling Pt: 265.4±25.0 °C (760 mm Hg)
Flash Pt: 114.3 °C
Storage Temperature: Ambient
MDL Number: MFCD20483545
CAS Number: 193290-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR