2-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Methoxycarbonylphenylboronic acid pinacol ester
A153392-250MG
A153392-1G
A153392-25G
A153392-5G
103804-506EA
10
USD
103804-506
103842-678
76783-472
77318-334
77318-336
77318-338
2-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
2-(Methoxycarbonyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BO₄ MW: 262.1093 g/mol Boiling Pt: 363.7 °C (760 mm Hg) Melting Pt: 49…51 °C Flash Pt: 173.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05663866 CAS Number: 653589-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...