4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-[(4-MethoxyCarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid methyll ester, Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, Methyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 4-(MethoxyCarbonylmethyl)phenylboronic acid Pinacol ester

A205194-10G A205194-250MG A205194-1G A205194-25G A205194-5G
103817-612EA 15 USD
103817-612 103848-278 76814-428 76814-430 77086-478 77086-480 77086-482
4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester 97%
4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
Boiling Pt: 363 °C (760 mm Hg)
Flash Pt: 173.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997797
CAS Number: 454185-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR