Methyl 2-(3-formylphenyl)acetate 95%

Supplier: Ambeed
Warning

A874171-100MG A874171-250MG A874171-1G
103799-736EA 66 USD
103799-736 103841-296 77019-810 77019-812 77308-476
Methyl 2-(3-formylphenyl)acetate 95%
Methyl 2-(3-formylphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15523582
CAS Number: 142327-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR