Methyl 2-(3-fluoro-4-nitrophenyl)acetate 95%

Supplier: Ambeed
Warning

A573338-250MG A573338-1G
103828-816EA 241 USD
103828-816 103839-958 77242-280 77307-852
Methyl 2-(3-fluoro-4-nitrophenyl)acetate 95%
Methyl 2-(3-fluoro-4-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈FNO₄
Storage Temperature: Ambient
MDL Number: MFCD16620330
CAS Number: 169339-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR