Methyl (3-bromophenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromophenylacetic acid methyll ester, Methyl 3-Bromophenylacetate, Methyl 2-(3-bromophenyl)acetate

A121943-100G A121943-10G A121943-25G A121943-500G A121943-1G A121943-5G
103800-156EA 6 USD
103800-156 77136-534 77136-536 77136-538 77136-540 77297-848 77312-288
Methyl (3-bromophenyl)acetate 98%
Methyl (3-bromophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.0700 g/mol
Boiling Pt: 268.6 °C (760 mm Hg)
Flash Pt: 116.3 °C
Storage Temperature: Ambient
MDL Number: MFCD06858764
CAS Number: 150529-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR