Methyl 2-(3-bromophenyl)-2-oxoacetate 95%

Supplier: Ambeed
Warning

A721225-1G A721225-100MG
103840-748EA 1118 USD
103840-748 76844-964 77018-858
Methyl 2-(3-bromophenyl)-2-oxoacetate 95%
Methyl 2-(3-bromophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11973930
CAS Number: 81316-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR