Methyl-2-(3-aminophenyl)acetate 97%

Supplier: Ambeed
Warning

A186904-250MG A186904-1G A186904-25G A186904-5G
103815-976EA 50 USD
103815-976 103846-564 76721-038 77302-052 77302-054 77302-056
Methyl-2-(3-aminophenyl)acetate 97%
Methyl-2-(3-aminophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
Boiling Pt: 281.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07366754
CAS Number: 52913-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR