Methyl 2-(3-amino-4-chlorophenyl)acetate 95%

Supplier: Ambeed
Warning

A155483-100MG A155483-250MG A155483-1G A155483-5G
103804-766EA 19 USD
103804-766 103842-964 76823-948 76823-950 77300-118 77300-120
Methyl 2-(3-amino-4-chlorophenyl)acetate 95%
Methyl 2-(3-amino-4-chlorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797914
CAS Number: 59833-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR