Methyl-2-(3-amino-2,6-difluorophenyl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(3-amino-2,6-difluorophenyl)acetate

A139022-10G A139022-250MG A139022-25G A139022-5G A139022-1G
103802-688EA 42 USD
103802-688 77272-394 77272-396 77299-042 77299-044 77315-762
Methyl-2-(3-amino-2,6-difluorophenyl)acetate 97%
Methyl-2-(3-amino-2,6-difluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
Boiling Pt: 267.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06658469
CAS Number: 361336-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR