3-(2-Methoxy-2-oxoethyl)phenylboronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

A168829-100MG A168829-250MG A168829-1G A168829-5G
103806-322EA 43 USD
103806-322 103844-576 76813-132 76813-134 77081-640 77081-642
3-(2-Methoxy-2-oxoethyl)phenylboronic acid, pinacol ester 95%
3-(2-Methoxy-2-oxoethyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
Boiling Pt: 372.043 °C (760 mm Hg)
Flash Pt: 178.806 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13195752
CAS Number: 478375-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR