Methyl (2-nitrophenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-nitrophenyl)acetate

A143816-100G A143816-10G A143816-1G A143816-25G A143816-5G
103803-320EA 6 USD
103803-320 77162-478 77162-480 77316-616 77316-618 77316-620
Methyl (2-nitrophenyl)acetate 98%
Methyl (2-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Boiling Pt: 283.4 °C (760 mm Hg)
Flash Pt: 126.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00968465
CAS Number: 30095-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR