Methyl (2-hydroxyphenyl)acetate 97%

Supplier: Ambeed
Warning

A143416-100G A143416-10G A143416-250MG A143416-25G A143416-5G
103803-276EA 12 USD
103803-276 77137-424 77137-426 77137-428 77299-340 77316-540
Methyl (2-hydroxyphenyl)acetate 97%
Methyl (2-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Boiling Pt: 261.9 °C (760 mm Hg)
Melting Pt: 122…124 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00090077
CAS Number: 22446-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR