Methyl-2-(2-formylphenyl)acetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-formylphenyl)acetate

A117350-100MG A117350-250MG A117350-1G
103798-870EA 95 USD
103798-870 103824-750 77026-208 77026-210 77297-534
Methyl-2-(2-formylphenyl)acetate 95%
Methyl-2-(2-formylphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.1900 g/mol
Boiling Pt: 273.9 °C (760 mm Hg)
Flash Pt: 117.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12755820
CAS Number: 63969-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR