Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95%

Supplier: Ambeed
Warning

A411255-100MG A411255-250MG A411255-1G A411255-5G
103827-222EA 11 USD
103827-222 103838-272 76829-348 76829-350 76841-806 76841-808
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95%
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Boiling Pt: 280.2±25.0 °C (760 mm Hg)
Flash Pt: 123.3 °C
Storage Temperature: Ambient
MDL Number: MFCD18399286
CAS Number: 91361-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR