Methyl 2-bromophenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-bromophenyl)acetate

A135881-100G A135881-10G A135881-1G A135881-25G A135881-5G
103802-236EA 5 USD
103802-236 76798-044 76798-046 77298-820 77298-822 77315-112
Methyl 2-bromophenylacetate 98%
Methyl 2-bromophenylacetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Boiling Pt: 264 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779430
CAS Number: 57486-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR