Methyl-1-methylpyrrole-2-acetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(1-methyl-1H-pyrrol-2-yl)acetate, 1-Methylpyrrole-2-acetic Acid Methyl Ester

A122999-100G A122999-10G A122999-5G
76971-654EA 441.34 USD
76971-654 76971-656 77312-442
Methyl-1-methylpyrrole-2-acetate 97%
Methyl-1-methylpyrrole-2-acetate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 242 °C (760 mm Hg)
Melting Pt: 210 °C (760 mm Hg)
Flash Pt: 88.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009752
CAS Number: 51856-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR