Methyl-2-(1H-indol-3-yl)-2-oxoacetate 97%

Supplier: Ambeed
Warning

A225489-1G A225489-25G A225489-5G A225489-100G A225489-10G
103819-184EA 31 USD
103819-184 103850-082 77088-780 77088-782 77088-784 77098-074 77098-076 77098-078
Methyl-2-(1H-indol-3-yl)-2-oxoacetate 97%
Methyl-2-(1H-indol-3-yl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
Boiling Pt: 383.2 °C (760 mm Hg)
Melting Pt: 227…230 °C
Flash Pt: 185.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00047173
CAS Number: 18372-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR