(±)-Boc-α-phosphonoglycine trimethyl ester 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate
A259384-25G
A259384-5G
A259384-100G
A259384-10G
A259384-1G
103812-440EA
9
USD
103812-440
103852-320
77091-880
77091-882
77113-654
77113-656
77304-766
(±)-Boc-α-phosphonoglycine trimethyl ester 98%
(±)-Boc-α-phosphonoglycine trimethyl ester
Precaución: for Research Use Only
Formula:
C₁₀H₂₀NO₇P MW: 297.24 g/mol Melting Pt: 66…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD02178893 CAS Number: 89524-98-1 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...