Methyl-2-((diphenylmethylene)amino)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-((diphenylmethylene)amino)acetate, N-(Diphenylmethylene)glycine methyl ester, Methyl N-(diphenylmethylene)glycinate

A169390-100G A169390-10G A169390-500G A169390-25G A169390-5G A169390-1000G
103844-636EA 5 USD
103844-636 77028-940 77028-942 77028-944 77081-708 77300-978 77667-364
Methyl-2-((diphenylmethylene)amino)acetate 97%
Methyl-2-((diphenylmethylene)amino)acetate

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₂
MW: 253.30 g/mol
Melting Pt: 42…44 °C
Flash Pt: 135.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272233
CAS Number: 81167-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR