(±)-Cbz-α-phosphonoglycine trimethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

A430918-100G A430918-10G A430918-1G A430918-25G A430918-5G
103827-516EA 6 USD
103827-516 103838-582 76725-384 76725-386 77307-252 77307-254 77307-256
(±)-Cbz-α-phosphonoglycine trimethyl ester 97%
(±)-Cbz-α-phosphonoglycine trimethyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
Melting Pt: 77…80 °C
Flash Pt: 244.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00043304
CAS Number: 88568-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR