Methyl-1-methyl-4-nitro-1H-pyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-4-nitro-1H-pyrazole-3-carboxylate

A169023-1G A169023-10G A169023-250MG
103806-342EA 16 USD
103806-342 103844-598 77081-668 77611-646 77683-384
Methyl-1-methyl-4-nitro-1H-pyrazole-3-carboxylate 98%
Methyl-1-methyl-4-nitro-1H-pyrazole-3-carboxylate

Precaución: For research use only.

Formula: C₆H₇N₃O₄
MW: 185.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19443825
CAS Number: 400877-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR