Methyl 1-methyl-1H-imidazole-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A142076-1G
A142076-5G
103803-092EA
11
USD
103803-092
77316-302
77316-304
Methyl 1-methyl-1H-imidazole-2-carboxylate 97%
Methyl 1-methyl-1H-imidazole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol Boiling Pt: 241.8 °C (760 mm Hg) Flash Pt: 100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179562 CAS Number: 62366-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...