Methyl-1-hydroxy-1-cyclopropane carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 1-hydroxycyclopropanecarboxylate
A133727-100G
A133727-10G
A133727-1G
A133727-25G
A133727-5G
A133727-250MG
103801-906EA
17
USD
103801-906
76782-500
76782-502
77314-586
77314-588
77314-590
77666-926
Methyl-1-hydroxy-1-cyclopropane carboxylate 95%
Methyl-1-hydroxy-1-cyclopropane carboxylate
Precaución: for Research Use Only
Formula:
C₅H₈O₃ MW: 116.116 g/mol Boiling Pt: 134.6 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD02093884 CAS Number: 33689-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...