Methyl 4-azaindole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate, 3-(Methoxycarbonyl)-1H-pyrrolo[3,2-b]pyridine

A181875-100MG A181875-250MG A181875-5G
103807-660EA 79 USD
103807-660 103846-032 76784-604 76784-606 77083-560
Methyl 4-azaindole-3-carboxylate 97%
Methyl 4-azaindole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864655
CAS Number: 952800-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR