Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-pyrrolo[2,3-b]pyridine-5-carboxylate

A174804-10G A174804-250MG A174804-1G
103806-976EA 23 USD
103806-976 103845-276 76985-602 76985-604 77301-352
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457785
CAS Number: 849067-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR