Methyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate

A186207-100MG A186207-250MG
103815-926EA 23 USD
103815-926 103846-502 76799-876 76799-878
Methyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate 97%
Methyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475793
CAS Number: 351439-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR