Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 7-azaindole-3-carboxylate, Methyl 1H-pyrrolo[2,3-b]pyridine-3-carboxylate

A118830-100MG A118830-250MG A118830-1G
103799-112EA 28 USD
103799-112 76810-348 76810-350 77297-626
Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylate 97%
Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864656
CAS Number: 808137-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR