Methyl-7-azaindole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate

A314457-1G A314457-25G A314457-5G A314457-10G
103825-034EA 30 USD
103825-034 103835-954 76838-766 76838-768 76838-770 77032-670
Methyl-7-azaindole-2-carboxylate 97%
Methyl-7-azaindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08752938
CAS Number: 394223-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR