Methyl-1H-pyrrole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-pyrrole-3-carboxylate

A109547-100G A109547-10G A109547-25G A109547-5G A109547-1G
103797-524EA 11 USD
103797-524 103823-324 77233-476 77233-478 77233-480 77283-872 77310-138
Methyl-1H-pyrrole-3-carboxylate 97%
Methyl-1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 284.1 °C (760 mm Hg)
Melting Pt: 85…89 °C
Flash Pt: 125.65 °C
Storage Temperature: Ambient
MDL Number: MFCD00967074
CAS Number: 2703-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR