Methyl-1H-pyrrole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl Pyrrole-2-carboxylate, Methyl 1H-pyrrole-2-carboxylate, Pyrrole-2-carboxylic acid methyll ester, Methyl pyrrole-2-carboxylate

A166723-100G A166723-10G A166723-1G A166723-500G A166723-25G A166723-5G
103844-356EA 10 USD
103844-356 77011-428 77011-430 77011-432 77011-434 77320-630 77320-632
Methyl-1H-pyrrole-2-carboxylate 98%
Methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂
Melting Pt: 74…78 °C
Flash Pt: 93.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00817048
CAS Number: 1193-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR