Methyl 1H-pyrazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A165507-100G
A165507-100MG
A165507-10G
A165507-250MG
A165507-1G
A165507-25G
A165507-5G
103805-972EA
10
USD
103805-972
103844-226
77096-282
77096-284
77096-286
77096-288
77320-436
77320-438
77320-440
Methyl 1H-pyrazole-4-carboxylate 97%
Methyl 1H-pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₅H₆N₂O₂ MW: 126.113 g/mol Boiling Pt: 271.1 °C (760 mm Hg) Melting Pt: 145…146 °C Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00297363 CAS Number: 51105-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...