Methyl-1H-pyrazole-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A242849-100G
A242849-10G
A242849-1G
A242849-250MG
A242849-25G
A242849-5G
103851-312EA
8
USD
103851-312
76801-378
76801-380
76801-382
76801-384
77090-502
77304-256
Methyl-1H-pyrazole-3-carboxylate 98%
Methyl-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₅H₆N₂O₂ Boiling Pt: 271.1 °C (760 mm Hg) Melting Pt: 141…142 °C Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00649381 CAS Number: 15366-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...