Methyl indole-6-carboxylate 98%

Supplier: Ambeed
Warning

A238051-100G A238051-10G A238051-500G A238051-1G A238051-25G A238051-5G
103811-200EA 11 USD
103811-200 103850-984 77031-218 77031-220 77031-222 77090-090 77090-092 77090-094
Methyl indole-6-carboxylate 98%
Methyl indole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Boiling Pt: 331.7 °C (760 mm Hg)
Melting Pt: 76…80 °C
Flash Pt: 154.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00211063
CAS Number: 50820-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR