Methyl indole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-indole-5-carboxylate

A114074-100G A114074-250MG A114074-10G A114074-25G
103798-340EA 16 USD
103798-340 103824-182 76796-994 76796-996 77565-998 77666-656
Methyl indole-5-carboxylate 98%
Methyl indole-5-carboxylate

Precaución: For research use only.

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 126…128 °C
Flash Pt: 154.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00153023
CAS Number: 1011-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR